CH4321848712 ChemBridge Corporation

7-(trifluoromethyl)-4-quinolinamine

  • MDL number MFCD00269411
  • Cas Number : 243666-11-7
  • Molecular Weight : 212.1712

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4043168

Safety Information

MSDS https://www.hit2lead.com/sds/4043168_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart