CH4321848715 ChemBridge Corporation

4-chloro-6-fluoro-2-methylquinoline

  • MDL number MFCD00272332
  • Cas Number : 18529-01-6
  • Molecular Weight : 195.6207

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4043187

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4043187
Availability details for
Quantity
  • Availability
  • Add to Cart