CH4321848726 ChemBridge Corporation

5-amino-2-hydroxy-3-methylbenzoic acid

  • MDL number MFCD04035023
  • Cas Number : 6265-14-1
  • Molecular Weight : 167.162

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4043368

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4043368
Availability details for
Quantity
  • Availability
  • Add to Cart