CH4321848733 ChemBridge Corporation

2-(2-aminophenyl)-2-propanol

  • MDL number MFCD04117984
  • Cas Number : 15833-00-8
  • Molecular Weight : 151.2056

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4043465

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4043465
Availability details for
Quantity
  • Availability
  • Add to Cart