CH4321848738 ChemBridge Corporation

3-chloro-5-methyl-1H-1,2,4-triazole

  • MDL number MFCD01451378
  • Cas Number : 15285-15-1
  • Molecular Weight : 117.537

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4018522

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4018522
Availability details for
Quantity
  • Availability
  • Add to Cart