CH4321848759 ChemBridge Corporation

4-methoxy-3-[(methylamino)carbonyl]benzenesulfonyl chloride

  • MDL number MFCD09039457
  • Cas Number : 918933-10-5
  • Molecular Weight : 263.698

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4043726

Safety Information

signalword Danger
hcodes 8
MSDS https://www.hit2lead.com/sds/4043726_sds.pdf
hstatements H314, H335.
Availability details for
Quantity
  • Availability
  • Add to Cart