CH4321848768 ChemBridge Corporation

1-(4,6-dichloro-5-pyrimidinyl)ethanone

  • MDL number MFCD13689148
  • Cas Number : 60025-06-1
  • Molecular Weight : 191.0148

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4043816

Safety Information

MSDS https://www.hit2lead.com/sds/4043816_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart