CH4321848771 ChemBridge Corporation

(3-methyl-2-pyrazinyl)methanol

  • MDL number MFCD09834939
  • Cas Number : 160818-32-6
  • Molecular Weight : 124.1405

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4036444

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4036444
Availability details for
Quantity
  • Availability
  • Add to Cart