CH4321848773 ChemBridge Corporation

5-chloro-2,3-dimethylpyrazine

  • MDL number MFCD13189517
  • Cas Number : 59489-32-6
  • Molecular Weight : 142.5862

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form liquid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4036448

Safety Information

MSDS https://www.hit2lead.com/sds/4036448_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart