CH4321848798 ChemBridge Corporation

4-chloro-1-methyl-1H-pyrrolo[2,3-b]pyridine

  • MDL number MFCD11501283
  • Cas Number : 74420-05-6
  • Molecular Weight : 166.6076

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4038240

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4038240
Availability details for
Quantity
  • Availability
  • Add to Cart