CH4321848825 ChemBridge Corporation

4,5-dimethyl-2-pyrimidinamine

  • MDL number MFCD00151982
  • Cas Number : 1193-74-4
  • Molecular Weight : 123.1558

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4028186

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4028186
Availability details for
Quantity
  • Availability
  • Add to Cart