CH4321848845 ChemBridge Corporation

5-(1H-pyrazol-1-yl)-2H-tetrazole

  • MDL number MFCD08848226
  • Cas Number : 1013759-52-8
  • Molecular Weight : 136.1148

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4044368

Safety Information

MSDS https://www.hit2lead.com/sds/4044368_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart