CH4321848855 ChemBridge Corporation

2,6-dimethyl-5-quinolinamine

  • MDL number MFCD00457007
  • Cas Number : 116632-59-8
  • Molecular Weight : 172.2264

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4100118

Safety Information

MSDS https://www.hit2lead.com/sds/4100118_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart