CH4321848901 ChemBridge Corporation

3,4,5-trimethoxybenzohydrazide

  • MDL number MFCD00014758
  • Cas Number : 3291-03-0
  • Molecular Weight : 226.2292

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4045167

Safety Information

MSDS https://www.hit2lead.com/sds/4045167_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart