CH4321848903 ChemBridge Corporation

3-(2-pyridinyl)-1H-pyrazole-5-carboxylic acid

  • MDL number MFCD05678901
  • Cas Number : 374064-02-5
  • Molecular Weight : 189.1708

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4401331

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4401331
Availability details for
Quantity
  • Availability
  • Add to Cart