CH4332968379 ChemBridge Corporation

2-(3-cyclopentyl-1-propyn-1-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

  • MDL number MFCD19982733
  • Cas Number : 1487355-69-0
  • Molecular Weight : 234.1422

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4044094

Safety Information

MSDS https://www.hit2lead.com/sds/4044094_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart