CH4332968385 ChemBridge Corporation

1-ethyl-6-oxo-1,6-dihydro-5-pyrimidinecarbonitrile

  • MDL number MFCD19982743
  • Cas Number : 1330756-12-1
  • Molecular Weight : 149.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4044342

Safety Information

MSDS https://www.hit2lead.com/sds/4044342_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart