CH4332968397 ChemBridge Corporation

5-isopropyl-1,3,4-thiadiazol-2-ol

  • MDL number MFCD19982758
  • Cas Number : 84352-67-0
  • Molecular Weight : 144.1948

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4044929

Safety Information

MSDS https://www.hit2lead.com/sds/4044929_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart