CH4332968515 ChemBridge Corporation

2-amino-6-butyl-5-methyl[1,2,4]triazolo[1,5-a]pyrimidin-7-ol

  • MDL number MFCD08691862
  • Cas Number : 873408-42-5
  • Molecular Weight : 221.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9040937

Safety Information

MSDS https://www.hit2lead.com/sds/9040937_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart