CH4332968524 ChemBridge Corporation

2-methyl-5-(methylsulfonyl)-4(3H)-pyrimidinone

  • MDL number MFCD20502922
  • Cas Number : 17551-48-3
  • Molecular Weight : 188.2043

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4044305

Safety Information

MSDS https://www.hit2lead.com/sds/4044305_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart