CH4332968530 ChemBridge Corporation

ethyl (6-oxo-1,6-dihydro-5-pyrimidinyl)acetate

  • MDL number MFCD11656636
  • Cas Number : 6214-46-6
  • Molecular Weight : 182.1766

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4044365

Safety Information

MSDS https://www.hit2lead.com/sds/4044365_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart