CH4332968542 ChemBridge Corporation

methyl 5-methyl-1H-imidazole-4-carboxylate

  • MDL number MFCD12964309
  • Cas Number : 78892-68-9
  • Molecular Weight : 140.1399

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4044514

Safety Information

MSDS https://www.hit2lead.com/sds/4044514_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart