CH4332968585 ChemBridge Corporation

1-(4-morpholinylsulfonyl)-1,4-diazepane

  • MDL number MFCD11168870
  • Cas Number : 756457-63-3
  • Molecular Weight : 249.3305

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form liquid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4044988

Safety Information

MSDS https://www.hit2lead.com/sds/4044988_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart