CH4332968586 ChemBridge Corporation

tert-butyl 4-(bromoacetyl)-1-piperazinecarboxylate

  • MDL number MFCD16990579
  • Cas Number : 112257-12-2
  • Molecular Weight : 307.1842

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4044994

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4044994
Availability details for
Quantity
  • Availability
  • Add to Cart