CH4358855841 ChemBridge Corporation

[4-(1-methyl-1H-pyrazol-5-yl)phenyl]boronic acid

  • MDL number MFCD21364389
  • Cas Number : 1487353-57-0
  • Molecular Weight : 202.0175

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. -20
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4045263

Safety Information

MSDS https://www.hit2lead.com/sds/4045263_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart