CH4358855843 ChemBridge Corporation

6-chloro-2,4-dimethylnicotinonitrile

  • MDL number MFCD21364391
  • Cas Number : 101251-72-3
  • Molecular Weight : 166.6076

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4045323

Safety Information

MSDS https://www.hit2lead.com/sds/4045323_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart