CH4358855896 ChemBridge Corporation

1-anilinocyclopentanecarboxylic acid

  • MDL number MFCD05864563
  • Cas Number : 6636-94-8
  • Molecular Weight : 205.253

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. -20C
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4140264

Safety Information

MSDS https://www.hit2lead.com/sds/4140264_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart