CH4358855909 ChemBridge Corporation

1-ethyl-3-methyl-1H-pyrazole-5-sulfonyl chloride

  • MDL number MFCD15976431
  • Cas Number : 1245820-78-3
  • Molecular Weight : 208.6659

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. -20
Physical Form liquid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4045430

Safety Information

signalword Danger
hcodes 8
MSDS https://www.hit2lead.com/sds/4045430_sds.pdf
hstatements H314, H335.
Availability details for
Quantity
  • Availability
  • Add to Cart