CH4358855932 ChemBridge Corporation

[(2S)-1-(4-methylbenzyl)-2-pyrrolidinyl]methanol

  • MDL number MFCD21364407
  • Cas Number : 87341-57-9
  • Molecular Weight : 205.2961

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4045576

Safety Information

MSDS https://www.hit2lead.com/sds/4045576_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart