CH4369259022 ChemBridge Corporation

5-(trifluoromethyl)-1,2,4-oxadiazol-3-amine

  • MDL number MFCD20720492
  • Cas Number : 883877-86-9
  • Molecular Weight : 153.0627

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4040449

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4040449
Availability details for
Quantity
  • Availability
  • Add to Cart