CH4369259024 ChemBridge Corporation

(1E)-N',2-dihydroxypropanimidamide

  • MDL number MFCD07776328
  • Cas Number : 52046-55-6
  • Molecular Weight : 104.1078

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4041720

Safety Information

MSDS https://www.hit2lead.com/sds/4041720_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart