CH4369259053 ChemBridge Corporation

2-methyl-8-quinolinesulfonyl chloride

  • MDL number MFCD17259588
  • Cas Number : 119373-30-7
  • Molecular Weight : 241.694

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4045420

Safety Information

signalword Danger
MSDS https://www.hit2lead.com/sds/4045420_sds.pdf
hstatements H314, H335.
Availability details for
Quantity
  • Availability
  • Add to Cart