CH4371462692 ChemBridge Corporation

3-bromo[1,2,4]triazolo[4,3-a]pyridine

  • MDL number MFCD11111699
  • Cas Number : 4922-68-3
  • Molecular Weight : 198.0201

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4045654

Safety Information

MSDS https://www.hit2lead.com/sds/4045654_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart