CH4371462718 ChemBridge Corporation

2-methoxypropanenitrile

  • MDL number MFCD16079402
  • Cas Number : 33695-59-9
  • Molecular Weight : 85.1045

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form liquid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4045776

Safety Information

MSDS https://www.hit2lead.com/sds/4045776_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart