CH4375444586 ChemBridge Corporation

2-hydrazino-4-(5-methyl-2-furyl)pyrimidine

  • MDL number MFCD22575205
  • Cas Number : 1410821-14-5
  • Molecular Weight : 190.2019

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4045126

Safety Information

MSDS https://www.hit2lead.com/sds/4045126_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart