CH4375444591 ChemBridge Corporation

4-(3-fluoro-4-methoxyphenyl)-2-hydrazinopyrimidine

  • MDL number MFCD22575212
  • Cas Number : 1410821-09-8
  • Molecular Weight : 234.2296

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4045187

Safety Information

MSDS https://www.hit2lead.com/sds/4045187_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart