CH4375444608 ChemBridge Corporation

2-chloro-4,6-dimethylnicotinic acid

  • MDL number MFCD02110082
  • Cas Number : 66662-48-4
  • Molecular Weight : 185.6076

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4042108

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4042108
Availability details for
Quantity
  • Availability
  • Add to Cart