CH4375444618 ChemBridge Corporation

5-methyl-4-phenyl-2-pyrimidinethiol

  • MDL number MFCD01689264
  • Cas Number : 857412-75-0
  • Molecular Weight : 202.2755

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4045060

Safety Information

signalword Warning
MSDS https://www.hit2lead.com/sds/4045060_sds.pdf
hstatements H302, H315, H319, H335.
Availability details for
Quantity
  • Availability
  • Add to Cart