CH4375444622 ChemBridge Corporation

2-hydrazino-4-(2-methoxyphenyl)pyrimidine

  • MDL number MFCD15204059
  • Cas Number : 1211451-37-4
  • Molecular Weight : 216.2392

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4045066

Safety Information

MSDS https://www.hit2lead.com/sds/4045066_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart