CH4375444626 ChemBridge Corporation

N-cyclopentyl-3-nitro-2-pyridinamine

  • MDL number MFCD08692157
  • Cas Number : 952934-74-6
  • Molecular Weight : 207.2291

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4045079

Safety Information

MSDS https://www.hit2lead.com/sds/4045079_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart