CH4375444640 ChemBridge Corporation

3-(2-fluorophenyl)-3-phenylpropanoic acid

  • MDL number MFCD12028377
  • Cas Number : 1142210-78-3
  • Molecular Weight : 244.2609

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4045104

Safety Information

MSDS https://www.hit2lead.com/sds/4045104_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart