CH4375444643 ChemBridge Corporation

5-bromo-N-(2-ethoxyethyl)-3-nitro-2-pyridinamine

  • MDL number MFCD14650450
  • Cas Number : 1249309-23-6
  • Molecular Weight : 290.1139

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4045113

Safety Information

MSDS https://www.hit2lead.com/sds/4045113_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart