CH4375444652 ChemBridge Corporation

4-(4-methoxyphenyl)-2-pyrimidinethiol

  • MDL number MFCD00085109
  • Cas Number : 175202-77-4
  • Molecular Weight : 218.2749

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4045145

Safety Information

signalword Warning
MSDS https://www.hit2lead.com/sds/4045145_sds.pdf
hstatements H302, H315, H319, H335.
Availability details for
Quantity
  • Availability
  • Add to Cart