CH4375444665 ChemBridge Corporation

2-ethoxy-2-methylpropanoic acid

  • MDL number MFCD14636712
  • Cas Number : 15001-71-5
  • Molecular Weight : 132.1577

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4021501

Safety Information

MSDS https://www.hit2lead.com/sds/4021501_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart