CH4375444669 ChemBridge Corporation

ethyl 5-cyano-6-ethoxy-2-methylnicotinate

  • MDL number MFCD05665984
  • Cas Number : 64119-44-4
  • Molecular Weight : 234.2512

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4045180

Safety Information

MSDS https://www.hit2lead.com/sds/4045180_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart