CH4375444670 ChemBridge Corporation

phenyl[4-(2-phenylethyl)phenyl]methanone

  • MDL number MFCD01076585
  • Cas Number : 91036-10-1
  • Molecular Weight : 286.367

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4045183

Safety Information

MSDS https://www.hit2lead.com/sds/4045183_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart