CH4375444676 ChemBridge Corporation

1-(benzyloxy)-4-bromo-2-nitrobenzene

  • MDL number MFCD13476870
  • Cas Number : 383868-64-2
  • Molecular Weight : 308.1274

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4045201

Safety Information

MSDS https://www.hit2lead.com/sds/4045201_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart