CH4375444680 ChemBridge Corporation

2-amino-5-bromobenzamide

  • MDL number MFCD04037881
  • Cas Number : 16313-66-9
  • Molecular Weight : 215.0473

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4045207

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4045207
Availability details for
Quantity
  • Availability
  • Add to Cart