CH4375444687 ChemBridge Corporation

2-amino-N-hydroxybutanamide

  • MDL number MFCD19625199
  • Cas Number : 36212-74-5
  • Molecular Weight : 118.1344

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4045655

Safety Information

MSDS https://www.hit2lead.com/sds/4045655_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart