CH4375444697 ChemBridge Corporation

3-ethyl-2(1H)-pyrazinone

  • MDL number MFCD19217676
  • Cas Number : 25680-54-0
  • Molecular Weight : 124.1405

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4045735

Safety Information

MSDS https://www.hit2lead.com/sds/4045735_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart